ENA341296451 Enamine

methyl[2-(thiophen-2-yl)ethyl]amine

  • MDL number MFCD04114520
  • Cas Number : 106891-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93489
Availability details for
Quantity
  • Availability
  • Add to Cart