ENA341296452 Enamine

1-[(thiophen-2-yl)methyl]-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD16681150
  • Cas Number : 1284940-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93490
Availability details for
Quantity
  • Availability
  • Add to Cart