ENA341296453 Enamine

3-[7-(difluoromethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl]propanoic acid

  • MDL number MFCD10001654
  • Cas Number : 1018053-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92470
Availability details for
Quantity
  • Availability
  • Add to Cart