ENA341296454 Enamine

4-(chloromethyl)-2-(methoxymethyl)-1,3-thiazole

  • MDL number MFCD11178189
  • Cas Number : 167155-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93492
Availability details for
Quantity
  • Availability
  • Add to Cart