ENA341296462 Enamine

4-bromo-5-methyl-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD08701232
  • Cas Number : 850832-55-2
  • Molecular Weight : 206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-92475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92475
Availability details for
Quantity
  • Availability
  • Add to Cart