ENA341296470 Enamine

methyl 1-phenylcyclopropane-1-carboxylate

  • MDL number MFCD12031860
  • Cas Number : 6121-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93514

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93514
Availability details for
Quantity
  • Availability
  • Add to Cart