ENA341296472 Enamine

2-{[(4-bromo-1H-pyrazol-1-yl)methyl]sulfanyl}acetic acid

  • MDL number MFCD05862135
  • Cas Number : 1006319-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92480
Availability details for
Quantity
  • Availability
  • Add to Cart