ENA341296475 Enamine

2-{[(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)methyl]sulfanyl}acetic acid

  • MDL number MFCD04970054
  • Cas Number : 1006436-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92481
Availability details for
Quantity
  • Availability
  • Add to Cart