ENA341296478 Enamine

[4-(3,5-dimethyl-1H-pyrazol-1-yl)phenyl]boronic acid

  • MDL number MFCD08572133
  • Cas Number : 1025495-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92482
Availability details for
Quantity
  • Availability
  • Add to Cart