ENA341296479 Enamine

4-bromo-1-methyl-3-nitro-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00464257
  • Cas Number : 84547-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92483
Availability details for
Quantity
  • Availability
  • Add to Cart