ENA341296480 Enamine

4-nitro-1-propyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04970194
  • Cas Number : 1006957-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92484
Availability details for
Quantity
  • Availability
  • Add to Cart