ENA341296485 Enamine

1-(4-nitrophenyl)piperidine-3-carboxylic acid

  • MDL number MFCD08442677
  • Cas Number : 926224-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92485
Availability details for
Quantity
  • Availability
  • Add to Cart