ENA341296487 Enamine

N-(3-fluoro-4-hydroxyphenyl)acetamide

  • MDL number MFCD18823529
  • Cas Number : 2045-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93567

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-93567
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart