ENA341296489 Enamine

1-[(4-chlorophenyl)methyl]-1H-pyrazol-4-amine

  • MDL number MFCD01459885
  • Cas Number : 97421-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93569

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93569
Availability details for
Quantity
  • Availability
  • Add to Cart