ENA341296491 Enamine

2-chloro-7-fluoroquinoline-6-sulfonyl chloride

  • MDL number MFCD11858140
  • Cas Number : 1118787-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93572
Availability details for
Quantity
  • Availability
  • Add to Cart