ENA341296496 Enamine

5-(1-chloroethyl)-3-(4H-1,2,4-triazol-3-yl)-1,2,4-oxadiazole

  • MDL number MFCD11203766
  • Cas Number : 1038358-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93578
Availability details for
Quantity
  • Availability
  • Add to Cart