ENA341296502 Enamine

4-methylmorpholine-2-carbonitrile

  • MDL number MFCD13190418
  • Cas Number : 1196152-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93591
Availability details for
Quantity
  • Availability
  • Add to Cart