ENA341296512 Enamine

3,4-dimethyl-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD03703466
  • Cas Number : 17825-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93601

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93601
Availability details for
Quantity
  • Availability
  • Add to Cart