ENA341296513 Enamine

4-chloro-1-(propan-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD06805223
  • Cas Number : 1006493-69-1
  • Molecular Weight : 188.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92489
Availability details for
Quantity
  • Availability
  • Add to Cart