ENA341296514 Enamine

6-bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD08276188
  • Cas Number : 108281-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93634

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93634
Availability details for
Quantity
  • Availability
  • Add to Cart