ENA341296540 Enamine

4-[2-(dimethylamino)ethoxy]benzene-1-carboximidamide

  • MDL number MFCD09944405
  • Cas Number : 1017049-06-7
  • Molecular Weight : 207.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93702
Availability details for
Quantity
  • Availability
  • Add to Cart