ENA341296541 Enamine

2-(4-methoxyphenyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD07375330
  • Cas Number : 884874-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93731
Availability details for
Quantity
  • Availability
  • Add to Cart