ENA341296557 Enamine

2-[4-(thiophen-3-ylmethyl)piperazin-1-yl]ethan-1-ol

  • MDL number MFCD00814072
  • Cas Number : 331858-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93749

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93749
Availability details for
Quantity
  • Availability
  • Add to Cart