ENA341296567 Enamine

3-methyl-6-oxo-1-(propan-2-yl)-1H,6H,7H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD12405247
  • Cas Number : 1160246-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92504
Availability details for
Quantity
  • Availability
  • Add to Cart