ENA341296577 Enamine

4-(4-bromo-3-cyclopropyl-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD06805424
  • Cas Number : 1006495-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92505
Availability details for
Quantity
  • Availability
  • Add to Cart