ENA341296580 Enamine

3-(4-amino-3,5-dimethyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD04969599
  • Cas Number : 1006486-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92506
Availability details for
Quantity
  • Availability
  • Add to Cart