ENA341296581 Enamine

6-methoxy-3,4-dihydro-2H-1,4-benzoxazine

  • MDL number MFCD08236770
  • Cas Number : 58960-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-59
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93833
Availability details for
Quantity
  • Availability
  • Add to Cart