ENA341296585 Enamine

1-cyclopropylcyclopropane-1-carboxylic acid

  • MDL number MFCD08704251
  • Cas Number : 60629-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93845
Availability details for
Quantity
  • Availability
  • Add to Cart