ENA341296588 Enamine

1-(2-methylphenyl)propan-2-one

  • MDL number MFCD00210465
  • Cas Number : 51052-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93852
Availability details for
Quantity
  • Availability
  • Add to Cart