ENA341296589 Enamine

1-azaspiro[3.3]heptan-2-one

  • MDL number MFCD20481993
  • Cas Number : 51047-68-8
  • Molecular Weight : 111.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-93854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93854
Availability details for
Quantity
  • Availability
  • Add to Cart