ENA341296590 Enamine

4-(1H-1,2,4-triazol-1-yl)butan-2-one

  • MDL number MFCD05863685
  • Cas Number : 884497-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93855
Availability details for
Quantity
  • Availability
  • Add to Cart