ENA341296591 Enamine

2-chloro-1-(chloromethyl)-4-methoxybenzene

  • MDL number MFCD12139149
  • Cas Number : 101080-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93856

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93856
Availability details for
Quantity
  • Availability
  • Add to Cart