ENA341296595 Enamine

3-[4-(methylthio)phenyl]-1,2,4-thiadiazol-5-amine

  • MDL number MFCD08436228
  • Cas Number : 889940-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93861

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93861
Availability details for
Quantity
  • Availability
  • Add to Cart