ENA341296598 Enamine

2-[4-(difluoromethyl)-3-methyl-6-oxo-1H,6H,7H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

  • MDL number MFCD10001634
  • Cas Number : 1018150-76-9
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92507
Availability details for
Quantity
  • Availability
  • Add to Cart