ENA341296603 Enamine

(2-aminoethoxy)trimethylsilane

  • MDL number MFCD01726954
  • Cas Number : 5804-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 24.9
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93871

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93871
Availability details for
Quantity
  • Availability
  • Add to Cart