ENA341296611 Enamine

3-[7-(trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl]propanoic acid

  • MDL number MFCD10001592
  • Cas Number : 1018170-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92511
Availability details for
Quantity
  • Availability
  • Add to Cart