ENA341296612 Enamine

2-(4-bromo-3-methyl-1H-pyrazol-1-yl)-2-methylpropanoic acid

  • MDL number MFCD04967802
  • Cas Number : 1006473-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92512

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92512
Availability details for
Quantity
  • Availability
  • Add to Cart