ENA341296621 Enamine

1-[(3-nitro-1H-pyrazol-1-yl)methyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06805378
  • Cas Number : 1006441-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92517
Availability details for
Quantity
  • Availability
  • Add to Cart