ENA341296625 Enamine

2-(2-bromo-4-formylphenoxy)acetic acid

  • MDL number MFCD02611621
  • Cas Number : 428838-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93914
Availability details for
Quantity
  • Availability
  • Add to Cart