ENA341296628 Enamine

1-[(4-methylphenyl)methyl]-5-oxopyrrolidine-2-carboxylic acid

  • MDL number MFCD03414586
  • Cas Number : 87341-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-93917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93917
Availability details for
Quantity
  • Availability
  • Add to Cart