ENA341296630 Enamine

1-(3-nitropyridin-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08696961
  • Cas Number : 1006442-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92519
Availability details for
Quantity
  • Availability
  • Add to Cart