ENA341296636 Enamine

2-(3-methylbutoxy)cyclopentan-1-amine

  • MDL number MFCD11126528
  • Cas Number : 1038313-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93926
Availability details for
Quantity
  • Availability
  • Add to Cart