ENA341296645 Enamine

3-chloro-4-(pyridazin-3-yloxy)aniline

  • MDL number MFCD16767765
  • Cas Number : 1249676-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93942
Availability details for
Quantity
  • Availability
  • Add to Cart