ENA341296649 Enamine

6-(trifluoromethoxy)-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD14654156
  • Cas Number : 1215279-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93948
Availability details for
Quantity
  • Availability
  • Add to Cart