ENA341296651 Enamine

2-{4H,6H,7H-pyrano[4,3-d][1,3]thiazol-2-yl}acetonitrile

  • MDL number MFCD06657758
  • Cas Number : 850349-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93950
Availability details for
Quantity
  • Availability
  • Add to Cart