ENA341296656 Enamine

6-chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine

  • MDL number MFCD11603432
  • Cas Number : 1082837-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93955
Availability details for
Quantity
  • Availability
  • Add to Cart