ENA341296657 Enamine

6-ethyl-3-methyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08754002
  • Cas Number : 937691-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92526
Availability details for
Quantity
  • Availability
  • Add to Cart