ENA341296658 Enamine

2-chloro-N-methoxy-N-methyl-5-nitrobenzene-1-sulfonamide

  • MDL number MFCD13207237
  • Cas Number : 1184485-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-93961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93961
Availability details for
Quantity
  • Availability
  • Add to Cart