ENA341296662 Enamine

2-{[(5-methyl-1,2-oxazol-3-yl)methyl]sulfanyl}acetic acid

  • MDL number MFCD09859430
  • Cas Number : 1018168-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92528
Availability details for
Quantity
  • Availability
  • Add to Cart