ENA341296664 Enamine

1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08696966
  • Cas Number : 1006962-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92530
Availability details for
Quantity
  • Availability
  • Add to Cart